missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethyl bromide, 98%, Thermo Scientific Chemicals

Product Code. 11454547
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in the synthesis and antihypertensive activity of 4'-substituted spiro[4H-3,1-benzoxazine-4,4'-piperidin]-2(1H)-ones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and antihypertensive activity of 4′-substituted spiro[4H-3,1-benzoxazine-4,4′-piperidin]-2(1H)-ones.

Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-10-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00000234
InChI Key JJFOBACUIRKUPN-UHFFFAOYSA-N
Synonym 2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene
PubChem CID 68526
IUPAC Name 2-bromoethoxybenzene
SMILES C1=CC=C(C=C1)OCCBr

Specifications

Melting Point 31°C to 35°C
Density 1.35
Boiling Point 135°C to 137°C (20 mmHg)
Flash Point 65°C (149°F)
Quantity 100 g
Beilstein 508290
Solubility Information Insoluble in water. Solubility in methanol is almost transparent. Soluble in chloroform and ethyl acetate.
Formula Weight 201.07
Percent Purity 98%
Chemical Name or Material 2-Phenoxyethyl bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'