missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethylamine, ≥95%, Thermo Scientific™

Product Code. 10171032
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1758-46-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
InChI Key IMLAIXAZMVDRGA-UHFFFAOYSA-N
Synonym 2-phenoxyethylamine, phenoxyethylamine, ethanamine, 2-phenoxy, 2-phenoxylethylamine, 2-phenoxy-ethylamine, 2-aminoethoxy benzene, ethylamine, 2-phenoxy, alpha-phenoxy-beta-aminoethane, unii-8dgq1b38r5, 2-phenoxyethylamino
PubChem CID 15651
IUPAC Name 2-phenoxyethanamine
SMILES C1=CC=C(C=C1)OCCN

Specifications

CAS Min % 95.0
Density 1.0480g/mL
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula C6H5O(CH2)2NH2
Quantity 1 g
Formula Weight 137.18
Percent Purity 95%
Chemical Name or Material 2-Phenoxyethylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'