missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-1,3-thiazole-4-carbonyl chloride, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 223.68 |
Percent Purity | 95% |
Chemical Name or Material | 2-Phenyl-1, 3-thiazole-4-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'