missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1-propylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11401520
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 280559-30-0
Molecular Formula C15H23BO2
Molecular Weight (g/mol) 246.157
MDL Number MFCD09953475
InChI Key GTZNZJQZUWIDTN-UHFFFAOYSA-N
Synonym 2-phenyl-1-propylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-phenylpropyl-1,3,2-dioxaborolane, 2-phenylpropylboronic acid pinacol ester, 2-2-phenylpropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 46739058
IUPAC Name 4,4,5,5-tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CC(C)C2=CC=CC=C2

Specifications

Quantity 5 g
Formula Weight 246.16
Percent Purity 97%
Chemical Name or Material 2-Phenyl-1-propylboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'