missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-pentenal, (E)+(Z), 90+%, Thermo Scientific Chemicals

Product Code. 11440214
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3491-63-2
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.216
MDL Number MFCD00051835
InChI Key YPAJRUMMODCONM-IZZDOVSWSA-N
Synonym 2-phenyl-2-pentenal, 2z-2-phenylpent-2-enal
PubChem CID 15310469
IUPAC Name (Z)-2-phenylpent-2-enal
SMILES CCC=C(C=O)C1=CC=CC=C1

Specifications

Density 1.005
Boiling Point 124°C to 126°C (14 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.551
Quantity 100 g
Beilstein 1859032
Sensitivity Air sensitive
Formula Weight 160.22
Percent Purity ≥90%
Chemical Name or Material 2-Phenyl-2-pentenal, (E)+(Z)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'