missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-2-propanol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.
Solubility
Insoluble in water.
Notes
Store away from acids, acid chlorides, acid anhydrides, oxidizing agents.
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.
Solubility
Insoluble in water.
Notes
Store away from acids, acid chlorides, acid anhydrides, oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 29°C to 34°C |
Density | 0.973 |
Boiling Point | 215°C |
Flash Point | 87°C (188°F) |
Refractive Index | 1.5196 |
Quantity | 25 g |
Beilstein | 1905012 |
Solubility Information | Insoluble in water. |
Formula Weight | 136.19 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'