missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylcyclohexanone, 98%, Thermo Scientific Chemicals

Product Code. 11493577
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It can be used to produce 6-oxo-6-phenyl-hexanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 6-oxo-6-phenyl-hexanoic acid.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1444-65-1
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.243
MDL Number MFCD00001629
InChI Key DRLVMOAWNVOSPE-UHFFFAOYSA-N
Synonym 2-phenylcyclohexanone, cyclohexanone, 2-phenyl, 2-phenyl-cyclohexanone, 2-phenyl cyclohexanone, cyclohexanone, phenyl, 2-phenylcyclohexanon, phenylcyclohexanone, phenyl cyclohexanone, 6-phenylcyclohexanone
PubChem CID 95592
IUPAC Name 2-phenylcyclohexan-1-one
SMILES C1CCC(=O)C(C1)C2=CC=CC=C2

Specifications

Melting Point 56°C to 59°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 174.24
Percent Purity 98%
Chemical Name or Material 2-Phenylcyclohexanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'