missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylphenol, 99%

Product Code. 11389599
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11389599

Brand: Thermo Scientific Alfa Aesar A10592.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 90-43-7 | C12H10O | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Phenylphenol is used as a dye intermediate, plasticizer, rubber chemical and as a preservative in water-oil emulsions. It is used as an antimicrobial preservative in cosmetics and an antimicrobial additive in the manufacture of metalworking fluids, leather, adhesives and textiles. Further, it serves as a disinfectant in households, hospitals, nursing homes, farms, laundries, barber shops, veterinary equipment and food processing plants. It is also employed in the production of resins and rubber chemicals. In addition to this, it is used as a polymer material, stabilizer and flame retardant.

Solubility
Soluble in ethanol, acetone, benzene,sodium hydroxide, chloroform, acetonitrile, toluene, hexane, ligroin, ethyl ether, pyridine, ethylene glycol, isopropanol, glycol ethers and polyglycols.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-43-7
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002208
InChI Key LLEMOWNGBBNAJR-UHFFFAOYSA-N
Synonym 2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
PubChem CID 7017
ChEBI CHEBI:17043
SMILES OC1=CC=CC=C1C1=CC=CC=C1

Specifications

Melting Point 54°C to 58°C
Density 1.293
Boiling Point 152°C to 154°C (15 mmHg)
Flash Point 124°C (255°F)
Odor Mild
Quantity 250 g
UN Number UN3077
Beilstein 606907
Merck Index 14,7304
Solubility Information Soluble in ethanol,acetone,benzene,sodium hydroxide,chloroform,acetonitrile,toluene,hexane,ligroin,ethyl ether,pyridine,ethylene glycol,isopropanol,glycol ethers and polyglycols.
IUPAC Name [1,1'-biphenyl]-2-ol
Formula Weight 170.21
Percent Purity 99%
Chemical Name or Material 2-Phenylphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Phenylphenol
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Phenylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.