missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylphenol, 99+%, Thermo Scientific Chemicals

Product Code. 13468348
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-43-7
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002208
InChI Key LLEMOWNGBBNAJR-UHFFFAOYSA-N
Synonym 2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
PubChem CID 7017
ChEBI CHEBI:17043
SMILES OC1=CC=CC=C1C1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point >56.0°C
Color White
Density 1.2100g/mL
Boiling Point 282.0°C
Flash Point 123°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5C6H4OH
Quantity 2.5 kg
Beilstein 06,II,623
Merck Index 15,7415
Specific Gravity 1.21
Solubility Information Solubility in water: 0.7g/L(20°C). Other solubilities: readily soluble in naoh solution,ethanol and acet,800g/L isopropanol,soluble in most common organic solvents
IUPAC Name 2-phenylphenol
Viscosity 2.4 mPa.s (100°C)
Formula Weight 170.21
Percent Purity 99+%
Physical Form Crystalline Flakes
Chemical Name or Material 2-Phenylphenol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Phenylphenol
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'