missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylphenol, 99+%, Thermo Scientific Chemicals

£16.78 - £165.83

Chemical Identifiers

CAS 90-43-7
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002208
InChI Key LLEMOWNGBBNAJR-UHFFFAOYSA-N
Synonym 2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
PubChem CID 7017
ChEBI CHEBI:17043
SMILES OC1=CC=CC=C1C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10427910
View Documents
Thermo Scientific Acros
130760050
5 g Glass bottle
£16.78
5g
Please to purchase this item. Need a web account? Register with us today!
10123280
View Documents Promotion Details
Thermo Scientific Acros
130765000
500 g Plastic bottle
£41.41
500g
Please to purchase this item. Need a web account? Register with us today!
13468348
View Documents
Thermo Scientific Acros
130760025
2.5 kg Plastic bottle
£165.83
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

90-43-7
170.21
LLEMOWNGBBNAJR-UHFFFAOYSA-N
7017
OC1=CC=CC=C1C1=CC=CC=C1
C12H10O
MFCD00002208
2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
CHEBI:17043

Specifications

90-43-7
100.0
White
282.0°C
Authentic
C12H10O
MFCD00002208
15,7415
2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
LLEMOWNGBBNAJR-UHFFFAOYSA-N
[1,1'-biphenyl]-2-ol
7017
2.4 mPa.s (100°C)
99+%
2-Phenylphenol, 99+%
99.0
>56.0°C
1.2100g/mL
123°C
99% min. (GC)
C6H5C6H4OH
06,II,623
1.21
Solubility in water: 0.7g/L(20°C). Other solubilities: readily soluble in naoh solution,ethanol and acet,800g/L isopropanol,soluble in most common organic solvents
OC1=CC=CC=C1C1=CC=CC=C1
170.21
CHEBI:17043
170.21
Crystalline Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 201-993-5

missing translation for 'rtecsNumber' : DV5775000

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Phenylphenol

Signal Word
  • Warning

Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates

RUO – Research Use Only