Learn More
(+/-)-2-Phenylpropionic acid, 98%, Thermo Scientific Chemicals
£18.90 - £199.00
Chemical Identifiers
CAS | 492-37-5 |
---|---|
Molecular Formula | C9H10O2 |
Molecular Weight (g/mol) | 150.177 |
MDL Number | MFCD00002650 |
InChI Key | YPGCWEMNNLXISK-UHFFFAOYSA-N |
Synonym | 2-phenylpropionic acid, hydratropic acid, dl-2-phenylpropionic acid, alpha-methylphenylacetic acid, 2-phenyl-propionic acid, +/--2-phenylpropionic acid, alpha-methylbenzeneacetic acid, benzeneacetic acid, .alpha.-methyl, hydratropasaeure, alpha-phenylpropionic acid |
PubChem CID | 10296 |
ChEBI | CHEBI:48526 |
IUPAC Name | 2-phenylpropanoic acid |
SMILES | CC(C1=CC=CC=C1)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11300184
|
Thermo Scientific Alfa Aesar
L00709.06 |
5 g |
£18.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11310184
|
Thermo Scientific Alfa Aesar
L00709.14 |
25 g |
£56.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320184
|
Thermo Scientific Alfa Aesar
L00709.22 |
100 g |
£199.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
492-37-5 | |
150.177 | |
YPGCWEMNNLXISK-UHFFFAOYSA-N | |
10296 | |
2-phenylpropanoic acid |
C9H10O2 | |
MFCD00002650 | |
2-phenylpropionic acid, hydratropic acid, dl-2-phenylpropionic acid, alpha-methylphenylacetic acid, 2-phenyl-propionic acid, +/--2-phenylpropionic acid, alpha-methylbenzeneacetic acid, benzeneacetic acid, .alpha.-methyl, hydratropasaeure, alpha-phenylpropionic acid | |
CHEBI:48526 | |
CC(C1=CC=CC=C1)C(=O)O |
Specifications
492-37-5 | |
1.1 | |
148°C (298°F) | |
1.523 | |
1863558 | |
YPGCWEMNNLXISK-UHFFFAOYSA-N | |
2-phenylpropanoic acid | |
10296 | |
150.18 | |
(+/-)-2-Phenylpropionic acid |
-5°C | |
264°C to 266°C | |
C9H10O2 | |
MFCD00002650 | |
2-phenylpropionic acid, hydratropic acid, dl-2-phenylpropionic acid, alpha-methylphenylacetic acid, 2-phenyl-propionic acid, +/--2-phenylpropionic acid, alpha-methylbenzeneacetic acid, benzeneacetic acid, .alpha.-methyl, hydratropasaeure, alpha-phenylpropionic acid | |
CC(C1=CC=CC=C1)C(=O)O | |
150.177 | |
CHEBI:48526 | |
98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 207-752-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only