Learn More
Tocris Bioscience™ 2-Phospho-L-ascorbic acid trisodium salt
Ascorbic acid derivative; maintains differentiation potential in bone marrow-derived MSCs
Brand: Tocris Bioscience™ 5778/50
32.83 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
2-Phospho-L-ascorbic acid trisodium salt is a stable ascorbic acid derivative used in cell culture; in combination with FGF-2, maintains differentiation potential in bone marrow-derived mesenchymal stem cells (MSC) through increased expression of HGF. Also exhibits synergistic protection of hMSCs under oxidative stress in combination with N-acetylcysteine (Cat. No. 5619).Specifications
2-(Dihydrogen phosphate)-L-ascorbic acid sodium salt | |
50 mg | |
C6H6Na3O9P | |
>95% |
66170-10-3 | |
Compounds for Stem Cell Differentiation | |
322.05 |