missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Picoline N-oxide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under dry inert gas. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates.
2-Picoline N-oxide is used in proteomics research. Also used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under dry inert gas. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates.
Specifications
Melting Point | 46°C to 50°C |
Boiling Point | 122°C (8 mmHg) |
Flash Point | 143°C (289°F) |
Quantity | 25 g |
Beilstein | 106916 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 109.13 |
Percent Purity | 98% |
Chemical Name or Material | 2-Picoline N-oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'