missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Piperidineethanol, 95%, Thermo Scientific Chemicals
£17.10 - £2101.09
Chemical Identifiers
CAS | 1484-84-0 |
---|---|
Molecular Formula | C7H15NO |
Molecular Weight (g/mol) | 129.2 |
InChI Key | PTHDBHDZSMGHKF-UHFFFAOYSA-N |
Synonym | 2-piperidineethanol, 2-2-hydroxyethyl piperidine, 2-piperidin-2-yl ethanol, 2-2-piperidyl ethan-1-ol, 2-piperidine ethanol, 2-hydroxyethyl piperidine, 2-piperidine-ethanol, piperidine-2-ethanol, 2-piperidin-2-yl ethan-1-ol |
PubChem CID | 15144 |
IUPAC Name | 2-piperidin-2-ylethanol |
SMILES | C1CCNC(C1)CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10474725
|
Thermo Scientific Acros
157610050 |
5 g | Glass bottle |
£17.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10163300
|
Thermo Scientific Acros
157611000 |
100 g | Glass bottle |
£148.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10031710
|
Thermo Scientific Acros
157615000 |
500 g | Glass bottle |
£566.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11946301
|
Thermo Scientific Acros
157610025 |
2.5 kg | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1484-84-0 | |
129.2 | |
2-piperidineethanol, 2-2-hydroxyethyl piperidine, 2-piperidin-2-yl ethanol, 2-2-piperidyl ethan-1-ol, 2-piperidine ethanol, 2-hydroxyethyl piperidine, 2-piperidine-ethanol, piperidine-2-ethanol, 2-piperidin-2-yl ethan-1-ol | |
2-piperidin-2-ylethanol |
C7H15NO | |
PTHDBHDZSMGHKF-UHFFFAOYSA-N | |
15144 | |
C1CCNC(C1)CCO |
Specifications
1484-84-0 | |
White to Yellow | |
234.0°C | |
Authentic | |
C7H15NO | |
1.01 | |
Solubility in water: very soluble. Other solubilities: soluble in alcohol | |
C1CCNC(C1)CCO | |
129.2 | |
129.2 | |
Crystalline Low Melting Solid |
38.0°C to 43.0°C | |
1.0100g/mL | |
102°C | |
94% min. (GC) | |
21,2 | |
2-piperidineethanol, 2-2-hydroxyethyl piperidine, 2-piperidin-2-yl ethanol, 2-2-piperidyl ethan-1-ol, 2-piperidine ethanol, 2-hydroxyethyl piperidine, 2-piperidine-ethanol, piperidine-2-ethanol, 2-piperidin-2-yl ethan-1-ol | |
PTHDBHDZSMGHKF-UHFFFAOYSA-N | |
2-piperidin-2-ylethanol | |
15144 | |
95% | |
2-Piperidineethanol |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 216-059-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Piperidineethanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only