Learn More
2-piperidino-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™
£74.80 - £184.00
Chemical Identifiers
CAS | 180403-13-8 |
---|---|
Molecular Formula | C9H12N2O2S |
Molecular Weight (g/mol) | 212.267 |
MDL Number | MFCD11506352 |
InChI Key | JIYHGZWZWANYAV-UHFFFAOYSA-N |
Synonym | 2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-piperidino-1,3-thiazole-5-carboxylic acid, 2-piperidyl-1,3-thiazole-5-carboxylic acid, 2-piperidin-1-yl thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2-1-piperidinyl |
PubChem CID | 33589513 |
IUPAC Name | 2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid |
SMILES | C1CCN(CC1)C2=NC=C(S2)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10326474
|
Thermo Scientific Maybridge
CC64101CB |
250 mg | Amber Glass Bottle |
£74.80
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10769015
|
Thermo Scientific Maybridge
CC64101DA |
1 g | Amber Glass Bottle |
£184.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
180403-13-8 | |
212.267 | |
JIYHGZWZWANYAV-UHFFFAOYSA-N | |
33589513 | |
C1CCN(CC1)C2=NC=C(S2)C(=O)O |
C9H12N2O2S | |
MFCD11506352 | |
2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-piperidino-1,3-thiazole-5-carboxylic acid, 2-piperidyl-1,3-thiazole-5-carboxylic acid, 2-piperidin-1-yl thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2-1-piperidinyl | |
2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid |
Specifications
180403-13-8 | |
100.0 | |
C9H12N2O2S | |
2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid, 2-piperidino-1,3-thiazole-5-carboxylic acid, 2-piperidyl-1,3-thiazole-5-carboxylic acid, 2-piperidin-1-yl thiazole-5-carboxylic acid, 5-thiazolecarboxylicacid, 2-1-piperidinyl | |
C1CCN(CC1)C2=NC=C(S2)C(=O)O | |
212.267 | |
212.27 | |
2-Piperidino-1, 3-thiazole-5-carboxylic acid |
96.5 | |
97% | |
MFCD11506352 | |
JIYHGZWZWANYAV-UHFFFAOYSA-N | |
2-piperidin-1-yl-1,3-thiazole-5-carboxylic acid | |
33589513 | |
97% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.