Learn More
2-piperidinobenzamide, 97%, Thermo Scientific™
£27.60 - £131.00
Chemical Identifiers
CAS | 3430-40-8 |
---|---|
Molecular Formula | C12H16N2O |
Molecular Weight (g/mol) | 204.27 |
MDL Number | MFCD00052257 |
InChI Key | VTXYPPVXMJMLCY-UHFFFAOYSA-N |
Synonym | 2-piperidinobenzamide, 2-piperidin-1-yl benzamide, benzamide,2-1-piperidinyl, 2-piperidylbenzamide, piperidinyl benzamide, maybridge1_000974, 2-1-piperidinyl benzamide, benzamide, 2-1-piperidinyl |
PubChem CID | 335067 |
SMILES | NC(=O)C1=CC=CC=C1N1CCCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10026082
|
Thermo Scientific Maybridge
BTB03853DA |
1 g |
£27.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10451913
|
Thermo Scientific Maybridge
BTB03853EA |
10 g |
£70.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10604575
|
Thermo Scientific Maybridge
|
25 g |
£131.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
3430-40-8 | |
204.27 | |
VTXYPPVXMJMLCY-UHFFFAOYSA-N | |
335067 |
C12H16N2O | |
MFCD00052257 | |
2-piperidinobenzamide, 2-piperidin-1-yl benzamide, benzamide,2-1-piperidinyl, 2-piperidylbenzamide, piperidinyl benzamide, maybridge1_000974, 2-1-piperidinyl benzamide, benzamide, 2-1-piperidinyl | |
NC(=O)C1=CC=CC=C1N1CCCCC1 |
Specifications
3430-40-8 | |
100.0 | |
Amber glass bottle | |
MFCD00052257 | |
VTXYPPVXMJMLCY-UHFFFAOYSA-N | |
2-piperidin-1-ylbenzamide | |
335067 | |
97% |
96.5 | |
97% | |
C12H16N2O | |
2-piperidinobenzamide, 2-piperidin-1-yl benzamide, benzamide,2-1-piperidinyl, 2-piperidylbenzamide, piperidinyl benzamide, maybridge1_000974, 2-1-piperidinyl benzamide, benzamide, 2-1-piperidinyl | |
NC(=O)C1=CC=CC=C1N1CCCCC1 | |
204.27 | |
204.27 | |
2-Piperidinobenzamide |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.