missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride, ≥95%, Thermo Scientific™

Catalog Number p-3075450
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
690634-81-2
C10H19ClN2O
218.73
MFCD06407992
RIAUQORBEIQABE-UHFFFAOYNA-N
2-pyrrolidine-1-carbonyl piperidine hydrochloride, 2-piperidinyl 1-pyrrolidinyl methanone hydrochloride, 2-pyrrolidin-1-yl carbonyl piperidine hydrochloride, piperidin-2-yl pyrrolidin-1-yl methanone hydrochloride, 1-2-piperidinylcarbonyl pyrrolidine hcl, 2-piperidyl pyrrolidinyl ketone, chloride, 2-pyrrolidin-1-ylcarbonyl piperidine hydrochloride
2794693
piperidin-2-yl(pyrrolidin-1-yl)methanone;hydrochloride
Cl.O=C(C1CCCCN1)N1CCCC1
This item is not returnable. View return policy
690634-81-2
C10H19ClN2O
218.73
MFCD06407992
RIAUQORBEIQABE-UHFFFAOYNA-N
2-pyrrolidine-1-carbonyl piperidine hydrochloride, 2-piperidinyl 1-pyrrolidinyl methanone hydrochloride, 2-pyrrolidin-1-yl carbonyl piperidine hydrochloride, piperidin-2-yl pyrrolidin-1-yl methanone hydrochloride, 1-2-piperidinylcarbonyl pyrrolidine hcl, 2-piperidyl pyrrolidinyl ketone, chloride, 2-pyrrolidin-1-ylcarbonyl piperidine hydrochloride
2794693
piperidin-2-yl(pyrrolidin-1-yl)methanone;hydrochloride
Cl.O=C(C1CCCCN1)N1CCCC1
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 218.73
Percent Purity 95%
Chemical Name or Material 2-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'