missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Piperidylmethyl)-1-cyclohexanone hydrochloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 166°C to 170°C |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C12H21NO·HCl |
Quantity | 1 g |
Merck Index | 13, 7510 |
Solubility Information | Solubility in water: soluble in water. Other solubilities: soluble in methanol,ethanol |
Molecular Weight (g/mol) | 231.77 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'