missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, For pesticide residue analysis, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Color | Colorless |
Density | 0.785g/mL at 25°C |
Quantity | 4 x 2.5 L |
Explosion Limit | 0.02 |
Flash Point | 12°C (53.6°F) |
Ignition Point | 750°F |
Linear Formula | (CH3)2CHOH |
Vapor Pressure | 33 mmHg (20°C), 44 mmHg (25°C) |
UN Number | UN1219 |
Formula Weight | 60.1 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'