missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, 99%, BAKER, J.T. Baker™

Product Code. 10200871
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
HDPE
Steel
This item is not returnable. View return policy
This item is not returnable. View return policy

For laboratory use, for organic synthesis, and for routine research.

A grade of chemicals for routine research usually controlled by physical criteria and often by assay (by GC).

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -88.5°C
Color Colorless
Acidity Max. 0.002%
Alkalinity Max. 0.002%
Density 0.781 to 0.783 g/mL at 25°C
Boiling Point 82.5°C
Quantity 2.5 L
Explosion Limit 2.5 - 12 Vol %
Flash Point 12°C
Ignition Point 399°C
Odor Alcohol
Packaging HDPE
Physical Form Liquid
Residue after Evaporation Max. 0.005%
Vapor Pressure 6.053 kPa at 25°C
Water Max. 0.2%
Formula Weight 60.1g/mol
Percent Purity ≥99%
Grade US FDA / ISO 9000 manufacturing, cGMP, IPEC member
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'