missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, BAKER HPLC ANALYZED, J.T. Baker™

Product Code. 15564055 Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
This item is not returnable. View return policy

Product Code. 15564055

Brand: JT Baker 9095.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

A group of high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations.

Specifications

pH Neutral
Melting Point -89.5°C
Absorbance At 254 nm, max. 0.02
CAS 67-63-0
Boiling Point 82°C
Quantity 2.5 L
Explosion Limit 2 to 12 vol%
Flash Point 12°C
Ignition Point 425°C
Odor Alcohol-like
Packaging Glass
Physical Form Liquid
Residue after Evaporation max. 2ppm
Molecular Formula C3H8O
UN Number 1219
Viscosity 2.43mP
Water max. 0.03 %
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Solubility Information Solubility in water at 20°C is 1g/l
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Percent Purity ≥99.7%
Grade HPLC
Chemical Name or Material 2-Propanol, HPLC
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-propanol
Signal Word
  • Danger||Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.