missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol CHROMASOLV™, for HPLC, 99.9%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
100 mL
185 L
2 L
2.5 L
2.5 L-PC
4 x 2.5 L
6 x 1 L
72 x 2.5 L
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
Melting Point | -89.5°C |
Density | 0.785g/mL at 25°C |
Boiling Point | 82°C |
Quantity | 2 L |
Refractive Index | n20/D 1.377 |
Linear Formula | (CH3)2CHOH |
Vapor Pressure | 44mmHg (25°C) |
Beilstein | 635639 |
Formula Weight | 60.1g/mol |
Percent Purity | 99.9% |
Show More |
Safety and Handling
Product Identifier
- 2-Propanol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction