missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, CHROMASOLV™ Plus, for HPLC, 99.9%, Honeywell Riedel-de Haën™
580.40 GBP valid until 2024-12-31
Use promo code "21958" to get your promotional price.
Use promo code "21958" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
4 x 2 L
4 x 2.5 L
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -89.5°C (literature) |
Absorbance | max. 0.005 at 400 nm, max. 0.005 at 275 nm, max. 0.30 at 250 nm, max. 1.0 at 245 nm |
CAS Max % | 100.0000% |
Linear Formula | (CH3)2CHOH |
UN Number | UN1219 |
Beilstein | 635639 |
Formula Weight | 60.11 |
Percent Purity | >99.85% |
Grade | HPLC |
Density | 0.785 g/mL at 25 °C(lit.) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'