missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, CHROMASOLV™ Plus, for HPLC, 99.9%, Honeywell Riedel-de Haën™

CHROMASOLV™ Plus, for HPLC, 99.9%

£72.75 - £720.34

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15636110
View Documents Promotion Details
Honeywell-Chromasolv
650447-1L
1 L
£72.75
1 L
Please to purchase this item. Need a web account? Register with us today!
15646110
View Documents
Honeywell-Chromasolv
650447-2.5L
2.5 L
£200.74
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15666110
View Documents
Honeywell-Chromasolv
650447-4X2L
4 x 2 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17199590
View Documents
Honeywell-Chromasolv
650447-4X2.5L
4 x 2.5 L
£720.34
7.86kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
Specifications

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O

Specifications

67-63-0
0.02
750°F
(CH3)2CHOH
33 mmHg (20°C), 44 mmHg (25°C)
635639
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1g/mol
CHROMASOLV™ Plus
0.785g/mL at 25°C
12°C (53.6°F)
C3H8O
MFCD00011674
UN1219
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
99%
2-Propanol
Videos
Safety and Handling

Safety and Handling

P210-P261-P305 + P351 + P338

missing translation for 'rtecsNumber' : NT8050000

Product Identifier
  • 2-Propanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications