missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, For pesticide residue analysis, Honeywell Riedel-de Haën™
CHROMASOLV™, for pesticide residue analysis
£22.46 - £322.10
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
MDL Number | MFCD00011674 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15674200
|
Honeywell-Riedel-de Haen
34486-1L |
1 L | N/A | ||||||
15684200
|
Honeywell-Riedel-de Haen
34486-2.5L |
2.5 L |
£92.57
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15694200
|
Honeywell-Riedel-de Haen
34486-4X2.5L |
4 x 2.5 L |
£322.10
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
MFCD00011674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Specifications
67-63-0 | |
0.785g/mL at 25°C | |
12°C (53.6°F) | |
C3H8O | |
MFCD00011674 | |
UN1219 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99.8% (GC) | |
2-Propanol |
Colorless | |
0.02 | |
750°F | |
(CH3)2CHOH | |
33 mmHg (20°C), 44 mmHg (25°C) | |
635639 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1g/mol | |
CHROMASOLV™ |
Safety and Handling
P210-P261-P305 + P351 + P338
missing translation for 'rtecsNumber' : NT8050000