missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, Laboratory Reagent, ≥99.5%, Honeywell™
Laboratory Reagent,≥99.5%
£29.39 - £375.46
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
MDL Number | MFCD00011674 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15650990
|
Honeywell Research Chemicals
109827-1L |
1 L | Glass Bottle | N/A | |||||
15660990
|
Honeywell Research Chemicals
109827-2.5L |
2.5 L | Glass Bottle |
£98.65
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15690990
|
Honeywell Research Chemicals
109827-4X2.5L |
4 x 2.5 L | Glass Bottle |
£375.46
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15640990
|
Honeywell Research Chemicals
109827-10L |
10 L | Stainless Steel Can | N/A | |||||
15670990
|
Honeywell Research Chemicals
109827-20L |
20 L | Stainless Steel Can | N/A | |||||
15680990
|
Honeywell Research Chemicals
109827-25L |
25 L | Stainless Steel Can | N/A | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
MFCD00011674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Specifications
-90°C | |
Colorless | |
77°C to 83°C (1013 hPa) | |
12°C (53.6°F) | |
Liquid | |
C3H8O | |
MFCD00011674 | |
UN1219 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1g/mol | |
Laboratory Reagent |
67-63-0 | |
0.785g/mL at 25°C | |
0.02 | |
750°F | |
n20/D 1.377(lit.) | |
(CH3)2CHOH | |
33mmHg (20°C), 44mmHg (25°C) | |
635639 | |
Soluble in water | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99.5% | |
2-Propanol |
Safety and Handling
P210-P261-P305 + P351 + P338
H225-H319-H336
missing translation for 'dotInformation' : Class 3 - PG 2 - Isopropanol
missing translation for 'einecsNumber' : 200-661-7
missing translation for 'rtecsNumber' : NT8050000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Propanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title