missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, puriss., ≥99.5% (GC), Honeywell Riedel-de Haën™
BIOPHARMA

Catalog Number 15692520
Click to view available options
Quantity:
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

Puriss., meets analytical specification of BP, Ph. Eur., USP,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Melting Point -85°C
Color Colorless
Density 0.785g/mL at 25°C
Boiling Point 82°C
Quantity 4 x 2.5 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Packaging Poly Bottle
Physical Form Liquid
Linear Formula (CH3)2CHOH
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
UN Number UN1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material 2-Propanol, Meets analytical specification of Ph. Eur.BP, USP
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'