missing translation for 'onlineSavingsMsg'
Learn More
2-Propanol, puriss., Honeywell Product Code.: 15652520

2-Propanol, puriss., Honeywell
BIOPHARMA

Product Code. 15652520
1 L
Click to view available options
Quantity:
100 mL
1 L
2.5 L
5 L
10 L
20 kg
25 L
160 kg
This item is not returnable. View return policy

Product Code. 15652520

Brand: Honeywell Riedel-de Haën 241371L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Density 0.785g/mL at 25°C
Quantity 1 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Linear Formula (CH3)2CHOH
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
UN Number 1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material 2-Propanol, Meets analytical specification of Ph. Eur.BP, USP
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN3380 Certificate TypeCertificate of Compliance Date27/02/2024 Catalog Number24137-1L
Lot NumberL3330 Certificate TypeCertificate of Analysis Date17/07/2023 Catalog Number24137-1L
Lot NumberN0550 Certificate TypeCertificate of Analysis Date05/04/2023 Catalog Number24137-4X5L,24137-5L,24137-6X1L,24137-160KG,24137-1L,24137-2.5L,24137-20KG,24137-25L,24137-4X2.5L
Lot NumberN0230 Certificate TypeCertificate of Analysis Date05/04/2023 Catalog Number24137-4X5L,24137-5L,24137-6X1L,24137-160KG,24137-1L,24137-2.5L,24137-20KG,24137-25L,24137-4X2.5L
Lot NumberM3460 Certificate TypeCertificate of Analysis Date05/04/2023 Catalog Number24137-4X5L,24137-5L,24137-6X1L,24137-160KG,24137-1L,24137-2.5L,24137-20KG,24137-25L,24137-4X2.5L
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Propanol, puriss., Honeywell >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.