missing translation for 'onlineSavingsMsg'
Learn More

2-Propylpentanoic acid, 99%

Product Code. 10246740 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10246740

Brand: Thermo Scientific Acros 208330250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-66-1 | C8H16O2 | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-66-1
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00002672
InChI Key NIJJYAXOARWZEE-UHFFFAOYSA-N
Synonym valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
PubChem CID 3121
ChEBI CHEBI:39867
IUPAC Name 2-propylpentanoic acid
SMILES CCCC(CCC)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 220.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CH2CH2)2CHCO2H
Refractive Index 1.4240 to 1.4260
Quantity 25 g
Beilstein 02, 350
Merck Index 15, 10099
Specific Gravity 0.92
Solubility Information Solubility in water: slightly soluble
Formula Weight 144.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Propylpentanoic acid, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Propylpentanoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.