Learn More
2-Propylpentanoic acid, sodium salt, 98%, Thermo Scientific Chemicals
£24.42 - £76.28
Chemical Identifiers
CAS | 1069-66-5 |
---|---|
MDL Number | MFCD00078604 |
InChI Key | AEQFSUDEHCCHBT-UHFFFAOYSA-M |
Synonym | sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerinShow More |
PubChem CID | 16760703 |
ChEBI | CHEBI:9925 |
IUPAC Name | sodium;2-propylpentanoate |
SMILES | CCCC(CCC)C(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10077682
|
Thermo Scientific Acros
271830050 |
5 g |
£24.42
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10466573
|
Thermo Scientific Acros
271830250 |
25 g |
£76.28
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1069-66-5 | |
AEQFSUDEHCCHBT-UHFFFAOYSA-M | |
16760703 | |
sodium;2-propylpentanoate |
MFCD00078604 | |
sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerinShow More | |
CHEBI:9925 | |
CCCC(CCC)C(=O)[O-].[Na+] |
Specifications
1069-66-5 | |
100.0 | |
White | |
98% | |
C8H15NaO2 | |
MFCD00078604 | |
sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin | |
AEQFSUDEHCCHBT-UHFFFAOYSA-M | |
sodium;2-propylpentanoate | |
16760703 | |
166.2 | |
Fine Powder |
97.5 | |
>300.0°C | |
Authentic | |
Glass bottle | |
(CH3CH2CH2)2CHCOONa | |
15, 10099 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
CCCC(CCC)C(=O)[O-].[Na+] | |
166.2 | |
CHEBI:9925 | |
98% | |
2-Propylpentanoic acid, sodium salt, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May damage the unborn child.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-961-8
missing translation for 'rtecsNumber' : YV7876000
missing translation for 'tsca' : TSCA
- 2-Propylpentanoic acid, sodium salt
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1A
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H360D-May damage the unborn child.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.