missing translation for 'onlineSavingsMsg'
Learn More
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals Product Code.: 10123330

2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals

Product Code. 10123330
100 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10123330

Brand: Thermo Scientific Acros 131711000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-97-5
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.1
MDL Number MFCD00006130
InChI Key NIPZZXUFJPQHNH-UHFFFAOYSA-N
Synonym 2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acid
PubChem CID 1047
ChEBI CHEBI:71311
IUPAC Name pyrazine-2-carboxylic acid
SMILES C1=CN=C(C=N1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 222.0°C to 225.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 25, 125
Merck Index 15, 8070
Solubility Information Solubility in water: soluble in cold water. Other solubilities: 1g/120g absolubte ethanol (25 c),practically insoluble in ether,chloroform and,benzene
Formula Weight 124.1
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Pyrazinecarboxylic acid, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435590 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number131711000,131710250
Lot NumberA0434539 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number131711000,131710250
Lot NumberA0440507 Certificate TypeCertificate of Analysis Date29/03/2022 Catalog Number131711000,131710250
Lot NumberA0421305 Certificate TypeCertificate of Analysis Date18/08/2020 Catalog Number131711000,131710250
Lot NumberA0359364 Certificate TypeCertificate of Analysis Date19/05/2020 Catalog Number131711000,131710250
Product Identifier
  • 2-Pyrazinecarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.