missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | >300°C |
Color | White |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 1 g |
IUPAC Name | 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane;propan-2-ol |
Formula Weight | 268.13 |
Percent Purity | 50-70%,see CofA for exact com |
Concentration or Composition (by Analyte or Components) | Contains varying amounts of IPA and N-phenyldiethanolamine |
Physical Form | Powder |
Show More |
Safety and Handling
Product Identifier
- 2-Pyridineboronic acid N-phenyldiethanolamine ester
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'