Learn More
2-Pyridinecarboxaldehyde, 99%, Thermo Scientific Chemicals
£29.80 - £202.00
Chemical Identifiers
CAS | 1121-60-4 |
---|---|
Molecular Formula | C6H5NO |
Molecular Weight (g/mol) | 107.11 |
MDL Number | MFCD00006290 |
InChI Key | CSDSSGBPEUDDEE-UHFFFAOYSA-N |
Synonym | 2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde |
PubChem CID | 14273 |
ChEBI | CHEBI:73012 |
IUPAC Name | pyridine-2-carbaldehyde |
SMILES | C1=CC=NC(=C1)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10755611
|
Thermo Scientific Acros
131820250 |
25 g | Glass bottle |
£29.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173420
|
Thermo Scientific Acros
131821000 |
100 g | Glass bottle |
£61.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10434405
|
Thermo Scientific Acros
131825000 |
500 g | Glass bottle |
£202.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1121-60-4 | |
107.11 | |
CSDSSGBPEUDDEE-UHFFFAOYSA-N | |
14273 | |
pyridine-2-carbaldehyde |
C6H5NO | |
MFCD00006290 | |
2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde | |
CHEBI:73012 | |
C1=CC=NC(=C1)C=O |
Specifications
1121-60-4 | |
100.0 | |
1.1210g/mL | |
73°C | |
99% | |
(C5H4N)COH | |
MFCD00006290 | |
1.121 | |
Solubility in water: miscible. Other solubilities: soluble in most common organic solvents | |
C1=CC=NC(=C1)C=O | |
107.11 | |
CHEBI:73012 | |
99% | |
2-Pyridinecarboxaldehyde |
98.5 | |
Brown-Yellow to Yellow | |
180.0°C to 182.0°C (761.0 mmHg) | |
Authentic | |
C6H5NO | |
1.5343 to 1.5373 | |
21, I, 287 | |
2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde | |
CSDSSGBPEUDDEE-UHFFFAOYSA-N | |
pyridine-2-carbaldehyde | |
14273 | |
107.11 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye damage.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occur
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 214-333-6
- 2-Pyridinecarboxaldehyde
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only