missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrimidinecarbonitrile, 98%, Thermo Scientific Chemicals

Product Code. 10582908
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14080-23-0
Molecular Formula C5H3N3
Molecular Weight (g/mol) 105.10
MDL Number MFCD00160513
InChI Key IIHQNAXFIODVDU-UHFFFAOYSA-N
Synonym 2-cyanopyrimidine, 2-pyrimidinecarbonitrile, 2-cyano-pyrimidine, 2-cyano pyrimidine, cyanopyrimidine, provastatin, pyrimidinecarbonitrile, zlchem 283, pubchem7042, pyrimidin-2-carbonitrile
PubChem CID 2757979
IUPAC Name pyrimidine-2-carbonitrile
SMILES N#CC1=NC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37.0°C to 41.0°C
Color White to Yellow
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 105.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Pyrimidinecarbonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2-Pyrimidinecarbonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'