missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-1-ylisonicotinic acid, ≥90%, Thermo Scientific™

Product Code. 10670454
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 98088-04-1
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD04115212
InChI Key PQLHQGBNMJTAEH-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-yl-isonicotinic acid, 2-pyrrolidin-1-yl isonicotinic acid, 2-pyrrolidin-1-yl pyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-1-pyrrolidinyl, 2-pyrrolidin-1-ylisonicotinic acid, 2-1-pyrrolidinyl isonicotinic acid, 2-1-pyrrolidinyl-4-pyridinecarboxylic acid, 2-pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate, 2-pyrrolidinylpyridine-4-carboxylic acid, hydrochloride
PubChem CID 6484236
IUPAC Name 2-pyrrolidin-1-ylpyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=NC=C1)N1CCCC1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 219.25
Percent Purity 90%
Chemical Name or Material 2-Pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'