Learn More
2-Pyrrolidin-2-ylpyridine, 95%, Thermo Scientific Chemicals
£178.00 - £731.00
Chemical Identifiers
CAS | 77790-61-5 |
---|---|
Molecular Formula | C9H12N2 |
Molecular Weight (g/mol) | 148.21 |
InChI Key | NDCZQFDBSPOUDF-UHFFFAOYSA-N |
Synonym | 2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine |
PubChem CID | 2771659 |
IUPAC Name | 2-pyrrolidin-2-ylpyridine |
SMILES | C1CC(NC1)C2=CC=CC=N2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13535000
|
Thermo Scientific Acros
446740010 |
1 g | Glass bottle |
£178.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13545000
|
Thermo Scientific Acros
446740050 |
5 g | Glass bottle |
£731.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
77790-61-5 | |
148.21 | |
2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine | |
2-pyrrolidin-2-ylpyridine |
C9H12N2 | |
NDCZQFDBSPOUDF-UHFFFAOYSA-N | |
2771659 | |
C1CC(NC1)C2=CC=CC=N2 |
Specifications
77790-61-5 | |
94% min. (GC) | |
2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine | |
C1CC(NC1)C2=CC=CC=N2 | |
148.21 | |
148.21 | |
2-Pyrrolidin-2-ylpyridine |
82.0°C (0.8 mmHg) | |
C9H12N2 | |
NDCZQFDBSPOUDF-UHFFFAOYSA-N | |
2-pyrrolidin-2-ylpyridine | |
2771659 | |
95% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
- (±)-2-(2-Pyrrolidinyl)pyridine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only