missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyrrolidinone, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 25.5°C |
Color | Colorless to Yellow |
Density | 1.1030g/mL |
Boiling Point | 250.0°C |
Flash Point | 138°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling

Product Identifier
- 2-Pyrrolidinone
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Pyrrolidinone, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction