missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidinone, 99%, Thermo Scientific Chemicals

£32.76 - £93.57

Chemical Identifiers

CAS 616-45-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00005270
InChI Key HNJBEVLQSNELDL-UHFFFAOYSA-N
Synonym 2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
PubChem CID 12025
ChEBI CHEBI:36592
IUPAC Name pyrrolidin-2-one
SMILES C1CC(=O)NC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11429150
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15332.36
500 g
£32.76
500g
Please to purchase this item. Need a web account? Register with us today!
11419150
View Documents
Thermo Scientific Alfa Aesar
A15332.A3
2 kg
£93.57
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.

Solubility
Miscible with water and other organic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
Specifications

Chemical Identifiers

616-45-5
85.106
HNJBEVLQSNELDL-UHFFFAOYSA-N
12025
pyrrolidin-2-one
C4H7NO
MFCD00005270
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
CHEBI:36592
C1CC(=O)NC1

Specifications

616-45-5
1.12
>110°C (230°F)
1.4875
Hygroscopic
2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
HNJBEVLQSNELDL-UHFFFAOYSA-N
pyrrolidin-2-one
12025
85.11
2-Pyrrolidinone
23°C to 25°C
245°C
C4H7NO
MFCD00005270
14,8016
Miscible with water and other organic solvents.
C1CC(=O)NC1
85.106
CHEBI:36592
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

missing translation for 'einecsNumber' : 210-483-1

missing translation for 'rtecsNumber' : UY5715000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Product Identifier
  • 2-Pyrrolidinone

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H319-Causes serious eye irritation.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 2-Pyrrolidinone

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.