missing translation for 'onlineSavingsMsg'
Learn More

2-Quinoxaloyl chloride, 95%, Thermo Scientific Chemicals

£49.10 - £151.00

Chemical Identifiers

CAS 54745-92-5
Molecular Formula C9H5ClN2O
Molecular Weight (g/mol) 192.6
InChI Key SOPDQKNXOCUBSR-UHFFFAOYSA-N
Synonym 2-quinoxalinecarbonyl chloride, 2-quinoxalinecarbonylchloride, 2-quinoxaloyl chloride, 2-quinoxaloylchloride, 2-quinoxalinylcarbonyl chloride, 2-quinoxaline carbonyl chloride, 2-quinoxalinecarbonyl chloride 7ci, 9ci
PubChem CID 2734681
IUPAC Name quinoxaline-2-carbonyl chloride
SMILES C1=CC=C2C(=C1)N=CC(=N2)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11946611
View Documents
Thermo Scientific Acros
163762500
250 mg
£49.10
250mg
Please to purchase this item. Need a web account? Register with us today!
Currently Unavailable: We have no information when this product will be available
View alternative products
11936611
View Documents
Thermo Scientific Acros
163760010
1 g
£151.00
1g
Please to purchase this item. Need a web account? Register with us today!
Currently Unavailable: We have no information when this product will be available
View alternative products
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54745-92-5
192.6
2-quinoxalinecarbonyl chloride, 2-quinoxalinecarbonylchloride, 2-quinoxaloyl chloride, 2-quinoxaloylchloride, 2-quinoxalinylcarbonyl chloride, 2-quinoxaline carbonyl chloride, 2-quinoxalinecarbonyl chloride 7ci, 9ci
quinoxaline-2-carbonyl chloride
C9H5ClN2O
SOPDQKNXOCUBSR-UHFFFAOYSA-N
2734681
C1=CC=C2C(=C1)N=CC(=N2)C(=O)Cl

Specifications

113.0°C to 115.0°C
Colorless to Yellow
100.0
94% min. (GC)
2-quinoxalinecarbonyl chloride, 2-quinoxalinecarbonylchloride, 2-quinoxaloyl chloride, 2-quinoxaloylchloride, 2-quinoxalinylcarbonyl chloride, 2-quinoxaline carbonyl chloride, 2-quinoxalinecarbonyl chloride 7ci, 9ci
C1=CC=C2C(=C1)N=CC(=N2)C(=O)Cl
192.6
192.6
Authentic
2-Quinoxaloyl chloride
54745-92-5
94.0
Liquid
C9H5ClN2O
SOPDQKNXOCUBSR-UHFFFAOYSA-N
quinoxaline-2-carbonyl chloride
2734681
95%
Glass Bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 259-315-9

Product Identifier
  • 2-Quinoxaloyl chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only