missing translation for 'onlineSavingsMsg'
Learn More

2-[(tert-Butoxycarbonyl)amino]-3-(3-fluorophenyl)propanoic acid, Tech., Thermo Scientific™

Catalog Number 10544905
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
114873-01-7
C14H18FNO4
283.299
MFCD00672522
FPCCREICRYPTTL-NSHDSACASA-N
boc-phe 3-f-oh, boc-l-3-fluorophenylalanine, boc-3-fluoro-l-phenylalanine, boc-l-phe 3-f-oh, s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, boc-l-3-fluorophe, boc-3-fluoro-l-phe, 2s-2-tert-butoxy carbonyl amino-3-3-fluorophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, n-boc-3-fluoro-l-phenylalanine
2734492
(2S)-3-(3-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)F)C(=O)O
This item is not returnable. View return policy
114873-01-7
C14H18FNO4
283.299
MFCD00672522
FPCCREICRYPTTL-NSHDSACASA-N
boc-phe 3-f-oh, boc-l-3-fluorophenylalanine, boc-3-fluoro-l-phenylalanine, boc-l-phe 3-f-oh, s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, boc-l-3-fluorophe, boc-3-fluoro-l-phe, 2s-2-tert-butoxy carbonyl amino-3-3-fluorophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, n-boc-3-fluoro-l-phenylalanine
2734492
(2S)-3-(3-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)F)C(=O)O
Assay Percent Range 90%
Quantity 10 g
Formula Weight 283.3
Percent Purity Tech.
Grade Technical
Chemical Name or Material (S)-N-(tert-Butoxycarbonyl)-3-fluorophenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'