missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(tert-butoxycarbonylamino)pyrimidine-5-boronic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 1032758-88-5 |
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C15H24BN3O4 |
MDL Number | MFCD09027069 |
Quantity | 1 g |
Synonym | 2-tert-butoxycarbonylamino pyrimidine-5-boronic acid, pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-yl carbamate, 2-tert-butoxycarbonylamino pyrimidine-5-boronic acid pinacol ester, tert-butyl n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-yl carbamate, tert-butyl n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-yl carbamate, 2-boc-amino pyrimidine-5-boronic acid pinacol ester, 2-tert-butoxycarbonylamino pyrimidine-5-boronic a, 2-tert-butoxycarbonyl amino pyrimidin-5-yl boronic acid pinacol ester, 2-tert-butoxycarbonylamino-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine |
InChI Key | XICHESWFGOSKMN-UHFFFAOYSA-N |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC(=O)OC(C)(C)C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'