missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Tetrahydropyran-4-yloxy)aniline, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 193.24 |
Percent Purity | 95% |
Chemical Name or Material | 2-(Tetrahydropyran-4-yloxy)aniline |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction