missing translation for 'onlineSavingsMsg'
Learn More

(2-Thien-2-ylpyrimidin-5-yl)methanol, 97%, Thermo Scientific™

Product Code. 10757675
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 921939-13-1
Molecular Formula C9H8N2OS
Molecular Weight (g/mol) 192.236
MDL Number MFCD09863233
InChI Key VPBKFPNQGGDRPA-UHFFFAOYSA-N
Synonym 2-thien-2-ylpyrimidin-5-yl methanol, 2-thiophen-2-yl pyrimidin-5-yl methanol, 2-thiophen-2-ylpyrimidin-5-yl methanol, 2-thien-2-yl pyrimidin-5-yl methanol, 5-hydroxymethyl-2-thien-2-ylpyrimidine, 2-2-thienyl pyrimidin-5-yl methan-1-ol, 2-thiophen-2-yl-pyrimidin-5-yl-methanol, 5-hydroxymethyl-2-thien-2-yl pyrimidine, 2-5-hydroxymethyl pyrimidin-2-yl thiophene
PubChem CID 24229731
IUPAC Name (2-thiophen-2-ylpyrimidin-5-yl)methanol
SMILES C1=CSC(=C1)C2=NC=C(C=N2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 192.24
Percent Purity 97%
Chemical Name or Material (2-Thien-2-ylpyrimidin-5-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'