missing translation for 'onlineSavingsMsg'
Learn More
2-Thienylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles Product Code.: 15475355

2-Thienylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15475355
50 mL
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy

Product Code. 15475355

Brand: Thermo Scientific Alfa Aesar H58397.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 45438-80-0 | C4H3BrSZn | 228.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thienylzinc bromide is used as medical intermediates.

Solubility
Reacts with water.

Notes
Air Sensitive. Store at 4°C. Incompatible with oxidizing agents, acid chlorides. Store under dry inert
TRUSTED_SUSTAINABILITY

Specifications

CAS 45438-80-0
Density 0.973
Molecular Formula C4H3BrSZn
MDL Number MFCD01317002
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym zinc 2h-thiophen-2-ide bromide, zinc 2+ ion thiophen-2-ide bromide
Solubility Information Reacts with water.
InChI Key KRDONBTWAGIZME-UHFFFAOYSA-M
SMILES [Zn+]Br.S1C=CC=[C-]1
IUPAC Name bromozinc(1+);2H-thiophen-2-ide
Molecular Weight (g/mol) 228.41
PubChem CID 4428324
Formula Weight 228.41
Chemical Name or Material 2-Thienylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ23F050 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH58397.AD
Lot NumberP24A028 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH58397.AD
Lot NumberT13D005 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH58397.AD
Lot NumberD20Y033 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH58397.AD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Thienylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.