missing translation for 'onlineSavingsMsg'
Learn More
2-Thiohydantoin, 98%, Thermo Scientific Chemicals Product Code.: 10193251

2-Thiohydantoin, 98%, Thermo Scientific Chemicals

Product Code. 10193251
25 g, Glass bottle
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10193251

Brand: Thermo Scientific Acros 138760250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 503-87-7
CAS Min % 98.5
CAS Max % 100.0
Melting Point 229.0°C to 231.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C3H4N2OS
MDL Number MFCD00005277
Quantity 25 g
Beilstein 24, 260
Synonym 2-thiohydantoin, thiohydantoin, 4-imidazolidinone, 2-thioxo, 2-thioxoimidazolidin-4-one, 2-thioguidanthion, hydantoin, 2-thio, usaf be-25, 2-thioxo-4-imidazolidinone, 2-thioxo-1,3-diazolidin-4-one, glycine thiohydantoin
Solubility Information Solubility in water: slightly.
InChI Key UGWULZWUXSCWPX-UHFFFAOYSA-N
SMILES O=C1CNC(=S)N1
IUPAC Name 2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol) 116.14
PubChem CID 1274030
Formula Weight 116.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiohydantoin, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Thiohydantoin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.