Learn More
2-Thiophenecarbonitrile, 97%, Thermo Scientific Chemicals
£31.30 - £426.00
Chemical Identifiers
CAS | 1003-31-2 |
---|---|
Molecular Formula | C5H3NS |
Molecular Weight (g/mol) | 109.15 |
MDL Number | MFCD00005416 |
InChI Key | CUPOOAWTRIURFT-UHFFFAOYSA-N |
Synonym | 2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile |
PubChem CID | 66087 |
IUPAC Name | thiophene-2-carbonitrile |
SMILES | C1=CSC(=C1)C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10702391
|
Thermo Scientific Acros
298050100 |
10 mL | Glass bottle |
£31.30
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10461681
|
Thermo Scientific Acros
298050500 |
50 mL | Glass bottle |
£110.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10125553
|
Thermo Scientific Acros
298052500 |
250 mL | Glass bottle |
£426.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1003-31-2 | |
109.15 | |
CUPOOAWTRIURFT-UHFFFAOYSA-N | |
66087 | |
C1=CSC(=C1)C#N |
C5H3NS | |
MFCD00005416 | |
2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile | |
thiophene-2-carbonitrile |
Specifications
1003-31-2 | |
100.0 | |
1.1700g/mL | |
53°C | |
96% min. (GC) | |
1.5620 to 1.564 | |
18,29 | |
2-thiophenecarbonitrile, 2-cyanothiophene, 2-thiophene carbonitrile, 2-thenylcyanide, thiophenecarbonitrile, 2-thienonitrile, 2-thenonitrile, 2-thiophenenitrile, 2-cyano-thiophene, 2-thienylcarbonitrile | |
C1=CSC(=C1)C#N | |
109.15 | |
109.15 | |
Liquid |
96.0 | |
Colorless to Yellow | |
192°C | |
Authentic | |
C5H3NS | |
MFCD00005416 | |
1.17 | |
CUPOOAWTRIURFT-UHFFFAOYSA-N | |
thiophene-2-carbonitrile | |
66087 | |
97% | |
2-Thiophenecarbonitrile, 97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-706-0
missing translation for 'tsca' : TSCA
RUO – Research Use Only