missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)pyridine, tech. 80%

Product Code. 11481387
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11481387

Brand: Thermo Scientific Alfa Aesar B24911.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17997-47-6 | C17H31NSn | 368.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17997-47-6
Molecular Formula C17H31NSn
Molecular Weight (g/mol) 368.152
MDL Number MFCD00052052
InChI Key GYUURHMITDQTRU-UHFFFAOYSA-N
Synonym 2-tributylstannyl pyridine, 2-tri-n-butylstannylpyridine, 2-tributylstannylpyridine, tributyl 2-pyridyl tin, 2-pyridyltributyltin, 2-1,1,1-tributylstannyl pyridine, 2-pyridyltri-n-butyltin, 2-tributylstannyl-pyridine, tri-n-butyl 2-pyridyl tin
PubChem CID 607784
IUPAC Name tributyl(pyridin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC=N1

Specifications

Density 1.15
Boiling Point 130°C to 132°C (0.8 mmHg)
Refractive Index 1.513
Quantity 1 g
UN Number UN2788
Beilstein 479329
Formula Weight 368.15
Percent Purity 80%
Grade Technical
Chemical Name or Material 2-(Tri-n-butylstannyl)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.