missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Tri-n-butylstannyl)thiazole, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Used as reagent for arylation of thiazole by Stille cross-coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as reagent for arylation of thiazole by Stille cross-coupling.
Solubility
Not miscible or difficult to mix.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Used as reagent for arylation of thiazole by Stille cross-coupling.
Solubility
Not miscible or difficult to mix.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.19 |
Boiling Point | 307°C to 309°C |
Quantity | 5 g |
UN Number | UN2788 |
Solubility Information | Not miscible or difficult to mix. |
Formula Weight | 374.17 |
Percent Purity | 96% |
Chemical Name or Material | 2-(Tri-n-butylstannyl)thiazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'