missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 10463685
Click to view available options
Quantity:
25 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1535-75-7
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00035959
InChI Key ZFCOUBUSGHLCDT-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy aniline, benzenamine, 2-trifluoromethoxy, 2-trifluoromethoxy-phenylamine, o-trifluoromethoxy aniline, alpha,alpha,alpha-trifluoro-o-anisidine, o-trifluoromethoxyaniline, 2-trifluoromethoxy aminobenzene, 2-trifluoromethoxy phenylamine, 2-trifluoromethoxy phenyl amine
PubChem CID 73754
IUPAC Name 2-(trifluoromethoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CF3OC6H4NH2
Quantity 50 g
Formula Weight 177.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'