missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific™
£23.68 - £51.52
Chemical Identifiers
CAS | 94651-33-9 |
---|---|
Molecular Formula | C8H5F3O2 |
Molecular Weight (g/mol) | 190.12 |
MDL Number | MFCD00042405 |
InChI Key | CPHXLFKIUVVIOQ-UHFFFAOYSA-N |
Synonym | 2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde |
PubChem CID | 2777192 |
IUPAC Name | 2-(trifluoromethoxy)benzaldehyde |
SMILES | FC(F)(F)OC1=CC=CC=C1C=O |
Description
Chemical Identifiers
94651-33-9 | |
190.12 | |
CPHXLFKIUVVIOQ-UHFFFAOYSA-N | |
2777192 | |
FC(F)(F)OC1=CC=CC=C1C=O |
C8H5F3O2 | |
MFCD00042405 | |
2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde | |
2-(trifluoromethoxy)benzaldehyde |
Specifications
94651-33-9 | |
100.0 | |
77°C (20.0 mmHg) | |
Authentic | |
Glass Bottle | |
MFCD00042405 | |
CPHXLFKIUVVIOQ-UHFFFAOYSA-N | |
2-(trifluoromethoxy)benzaldehyde | |
2777192 | |
96% | |
2-(Trifluoromethoxy)benzaldehyde |
95.0 | |
Colorless to Yellow | |
67°C | |
95% min. (GC) | |
C8H5F3O2 | |
2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde | |
FC(F)(F)OC1=CC=CC=C1C=O | |
190.12 | |
190.12 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific™